Assay Details
Gene Symbol: LOC5567142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651489.2 | XP_001651539.2 | 1 - 2 | 527 | 63 | |
XM_021841088.1 | XP_021696780.1 | 1 - 2 | 531 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001651489.2, XM_021841088.1 transcripts.
Set Membership: |
![]() ![]() |
