Assay Details
Gene Symbol: LOC5567142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651489.2 | XP_001651539.2 | 4 - 5 | 1410 | 80 | |
XM_021841087.1 | XP_021696779.1 | 4 - 5 | 1056 | 80 | ||
XM_021841088.1 | XP_021696780.1 | 4 - 5 | 1414 | 80 |
