Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 9 - 10 | 3542 | 67 | |
XM_021854765.1 | XP_021710457.1 | 8 - 9 | 3516 | 67 | ||
XM_021854766.1 | XP_021710458.1 | 9 - 10 | 3499 | 67 | ||
XM_021854767.1 | XP_021710459.1 | 9 - 10 | 3590 | 67 | ||
XM_021854768.1 | XP_021710460.1 | 8 - 9 | 3473 | 67 | ||
XM_021854769.1 | XP_021710461.1 | 8 - 9 | 3564 | 67 | ||
XM_021854770.1 | XP_021710462.1 | 9 - 10 | 3296 | 67 | ||
XM_021854771.1 | XP_021710463.1 | 9 - 10 | 3362 | 67 | ||
XM_021854772.1 | XP_021710464.1 | 8 - 9 | 3263 | 67 | ||
XM_021854773.1 | XP_021710465.1 | 8 - 9 | 3338 | 67 |

More Information
Set Membership: |
![]() ![]() |
