Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 8 - 9 | 3200 | 86 | |
XM_021854765.1 | XP_021710457.1 | 7 - 8 | 3174 | 86 | ||
XM_021854766.1 | XP_021710458.1 | 8 - 9 | 3157 | 86 | ||
XM_021854767.1 | XP_021710459.1 | 8 - 9 | 3248 | 86 | ||
XM_021854768.1 | XP_021710460.1 | 7 - 8 | 3131 | 86 | ||
XM_021854769.1 | XP_021710461.1 | 7 - 8 | 3222 | 86 | ||
XM_021854770.1 | XP_021710462.1 | 8 - 9 | 2954 | 86 | ||
XM_021854771.1 | XP_021710463.1 | 8 - 9 | 3020 | 86 | ||
XM_021854772.1 | XP_021710464.1 | 7 - 8 | 2921 | 86 | ||
XM_021854773.1 | XP_021710465.1 | 7 - 8 | 2996 | 86 |
