Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 6 - 7 | 2678 | 84 | |
XM_021854765.1 | XP_021710457.1 | 5 - 6 | 2652 | 84 | ||
XM_021854766.1 | XP_021710458.1 | 6 - 7 | 2635 | 84 | ||
XM_021854767.1 | XP_021710459.1 | 6 - 7 | 2726 | 84 | ||
XM_021854768.1 | XP_021710460.1 | 5 - 6 | 2609 | 84 | ||
XM_021854769.1 | XP_021710461.1 | 5 - 6 | 2700 | 84 | ||
XM_021854770.1 | XP_021710462.1 | 6 - 7 | 2432 | 84 | ||
XM_021854771.1 | XP_021710463.1 | 6 - 7 | 2498 | 84 | ||
XM_021854772.1 | XP_021710464.1 | 5 - 6 | 2399 | 84 | ||
XM_021854773.1 | XP_021710465.1 | 5 - 6 | 2474 | 84 |
