Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 5 - 6 | 2510 | 82 | |
XM_021854765.1 | XP_021710457.1 | 4 - 5 | 2484 | 82 | ||
XM_021854766.1 | XP_021710458.1 | 5 - 6 | 2467 | 82 | ||
XM_021854767.1 | XP_021710459.1 | 5 - 6 | 2558 | 82 | ||
XM_021854768.1 | XP_021710460.1 | 4 - 5 | 2441 | 82 | ||
XM_021854769.1 | XP_021710461.1 | 4 - 5 | 2532 | 82 | ||
XM_021854770.1 | XP_021710462.1 | 5 - 6 | 2264 | 82 | ||
XM_021854771.1 | XP_021710463.1 | 5 - 6 | 2330 | 82 | ||
XM_021854772.1 | XP_021710464.1 | 4 - 5 | 2231 | 82 | ||
XM_021854773.1 | XP_021710465.1 | 4 - 5 | 2306 | 82 |
