Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 11 - 12 | 3774 | 81 | |
XM_021837332.1 | XP_021693024.1 | 11 - 12 | 3816 | 81 | ||
XM_021837333.1 | XP_021693025.1 | 11 - 12 | 3907 | 81 | ||
XM_021837334.1 | XP_021693026.1 | 10 - 11 | 4155 | 81 | ||
XM_021837335.1 | XP_021693027.1 | 12 - 13 | 3813 | 81 |
