Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 12 - 13 | 4225 | 67 | |
XM_021837332.1 | XP_021693024.1 | 12 - 13 | 4267 | 67 | ||
XM_021837333.1 | XP_021693025.1 | 12 - 13 | 4358 | 67 | ||
XM_021837334.1 | XP_021693026.1 | 11 - 12 | 4606 | 67 | ||
XM_021837335.1 | XP_021693027.1 | 13 - 14 | 4264 | 67 |

More Information
Set Membership: |
![]() ![]() |
