Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 15 - 16 | 5047 | 104 | |
XM_021837332.1 | XP_021693024.1 | 15 - 16 | 5089 | 104 | ||
XM_021837333.1 | XP_021693025.1 | 15 - 16 | 5180 | 104 | ||
XM_021837334.1 | XP_021693026.1 | 14 - 15 | 5428 | 104 | ||
XM_021837335.1 | XP_021693027.1 | 16 - 17 | 5086 | 104 |

More Information
Set Membership: |
![]() ![]() |
