Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 18 - 19 | 5439 | 71 | |
XM_021837332.1 | XP_021693024.1 | 18 - 19 | 5481 | 71 | ||
XM_021837333.1 | XP_021693025.1 | 18 - 19 | 5572 | 71 | ||
XM_021837334.1 | XP_021693026.1 | 17 - 18 | 5820 | 71 | ||
XM_021837335.1 | XP_021693027.1 | 19 - 20 | 5478 | 71 |
