Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 1 - 2 | 540 | 59 | |
XM_021837335.1 | XP_021693027.1 | 1 - 2 | 540 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021837330.1, XM_021837335.1 transcripts.
Set Membership: |
![]() ![]() |
