Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 21 - 22 | 6853 | 101 | |
XM_021837332.1 | XP_021693024.1 | 21 - 22 | 6895 | 101 | ||
XM_021837333.1 | XP_021693025.1 | 21 - 22 | 6986 | 101 | ||
XM_021837334.1 | XP_021693026.1 | 20 - 21 | 7234 | 101 | ||
XM_021837335.1 | XP_021693027.1 | 22 - 23 | 6892 | 101 |

More Information
Set Membership: |
![]() ![]() |
