Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 5 - 6 | 1495 | 98 | |
XM_021837332.1 | XP_021693024.1 | 5 - 6 | 1537 | 98 | ||
XM_021837333.1 | XP_021693025.1 | 5 - 6 | 1628 | 98 | ||
XM_021837334.1 | XP_021693026.1 | 4 - 5 | 1876 | 98 | ||
XM_021837335.1 | XP_021693027.1 | 6 - 7 | 1534 | 98 |
