Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 7 - 8 | 2094 | 115 | |
XM_021837332.1 | XP_021693024.1 | 7 - 8 | 2136 | 115 | ||
XM_021837333.1 | XP_021693025.1 | 7 - 8 | 2227 | 115 | ||
XM_021837334.1 | XP_021693026.1 | 6 - 7 | 2475 | 115 | ||
XM_021837335.1 | XP_021693027.1 | 8 - 9 | 2133 | 115 |

More Information
Set Membership: |
![]() ![]() |
