Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 8 - 9 | 3462 | 87 | |
XM_021837332.1 | XP_021693024.1 | 8 - 9 | 3504 | 87 | ||
XM_021837333.1 | XP_021693025.1 | 8 - 9 | 3595 | 87 | ||
XM_021837334.1 | XP_021693026.1 | 7 - 8 | 3843 | 87 | ||
XM_021837335.1 | XP_021693027.1 | 9 - 10 | 3501 | 87 |
