Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 10 - 11 | 3711 | 98 | |
XM_021837332.1 | XP_021693024.1 | 10 - 11 | 3753 | 98 | ||
XM_021837333.1 | XP_021693025.1 | 10 - 11 | 3844 | 98 | ||
XM_021837334.1 | XP_021693026.1 | 9 - 10 | 4092 | 98 | ||
XM_021837335.1 | XP_021693027.1 | 11 - 12 | 3750 | 98 |
