Assay Details
Gene Symbol: LOC5565069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857635.1 | XP_021713327.1 | 3 - 4 | 1359 | 68 | |
XM_021857636.1 | XP_021713328.1 | 2 - 3 | 2815 | 68 |

More Information
Set Membership: |
![]() ![]() |
