Assay Details
Gene Symbol: LOC5565069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857635.1 | XP_021713327.1 | 6 - 7 | 2697 | 58 | |
XM_021857636.1 | XP_021713328.1 | 5 - 6 | 4153 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
