Assay Details
Gene Symbol: LOC5577722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844556.1 | XP_021700248.1 | 7 - 8 | 1630 | 74 | |
XM_021844557.1 | XP_021700249.1 | 7 - 8 | 1630 | 74 | ||
XM_021844558.1 | XP_021700250.1 | 6 - 7 | 1492 | 74 | ||
XM_021844559.1 | XP_021700251.1 | 7 - 8 | 1630 | 74 |
