Assay Details
Gene Symbol: LOC5577722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844556.1 | XP_021700248.1 | 16 - 17 | 3669 | 72 | |
XM_021844557.1 | XP_021700249.1 | 16 - 17 | 3561 | 72 | ||
XM_021844558.1 | XP_021700250.1 | 15 - 16 | 3534 | 72 | ||
XM_021844559.1 | XP_021700251.1 | 15 - 16 | 3378 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
