Assay Details
Gene Symbol: LOC5577722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844556.1 | XP_021700248.1 | 15 - 16 | 3553 | 69 | |
XM_021844558.1 | XP_021700250.1 | 14 - 15 | 3418 | 69 |

More Information
Set Membership: |
![]() ![]() |
