Assay Details
Gene Symbol: LOC5577722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844556.1 | XP_021700248.1 | 14 - 15 | 3263 | 72 | |
XM_021844557.1 | XP_021700249.1 | 14 - 15 | 3266 | 72 | ||
XM_021844558.1 | XP_021700250.1 | 13 - 14 | 3128 | 72 |
