Assay Details
Gene Symbol: LOC5577722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844556.1 | XP_021700248.1 | 11 - 12 | 2720 | 57 | |
XM_021844557.1 | XP_021700249.1 | 11 - 12 | 2720 | 57 | ||
XM_021844558.1 | XP_021700250.1 | 10 - 11 | 2582 | 57 | ||
XM_021844559.1 | XP_021700251.1 | 11 - 12 | 2720 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
