Assay Details
Gene Symbol: LOC5571215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845773.1 | XP_021701465.1 | 1 - 2 | 659 | 84 | |
GenBank mRNA | GQ231545.1 | - | 1 - 2 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845773.1 transcripts.
Set Membership: |
![]() ![]() |
