Assay Details
Gene Symbol: LOC5571215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845773.1 | XP_021701465.1 | 6 - 7 | 1861 | 88 | |
GenBank mRNA | GQ231545.1 | - | 6 - 7 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
