Assay Details
Gene Symbol: LOC5571798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854009.1 | XP_021709701.1 | 13 - 14 | 1923 | 65 | |
XM_021854066.1 | XP_021709758.1 | 13 - 14 | 1920 | 65 | ||
XM_021854131.1 | XP_021709823.1 | 13 - 14 | 1908 | 65 | ||
XM_021854182.1 | XP_021709874.1 | 13 - 14 | 1905 | 65 | ||
XM_021854254.1 | XP_021709946.1 | 12 - 13 | 1866 | 65 | ||
XM_021854314.1 | XP_021710006.1 | 12 - 13 | 1851 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
