Assay Details
Gene Symbol: LOC5566179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839137.1 | XP_021694829.1 | 1 - 2 | 132 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839137.1 transcripts.
Set Membership: |
![]() ![]() |
