Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 12 - 13 | 4288 | 86 | |
XM_021850418.1 | XP_021706110.1 | 12 - 13 | 4287 | 86 | ||
XM_021850419.1 | XP_021706111.1 | 8 - 9 | 2780 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
