Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 8 - 9 | 2051 | 78 | |
XM_021850418.1 | XP_021706110.1 | 8 - 9 | 2050 | 78 | ||
XM_021850419.1 | XP_021706111.1 | 4 - 5 | 543 | 78 |
