Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 4 - 5 | 907 | 61 | |
XM_021844764.1 | XP_021700456.1 | 4 - 5 | 1019 | 61 | ||
XM_021844765.1 | XP_021700457.1 | 4 - 5 | 863 | 61 | ||
XM_021844766.1 | XP_021700458.1 | 4 - 5 | 964 | 61 | ||
XM_021844767.1 | XP_021700459.1 | 4 - 5 | 850 | 61 | ||
XM_021844768.1 | XP_021700460.1 | 3 - 4 | 735 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
