Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 2 - 3 | 266 | 79 | |
XM_021844764.1 | XP_021700456.1 | 2 - 3 | 378 | 79 | ||
XM_021844765.1 | XP_021700457.1 | 2 - 3 | 222 | 79 | ||
XM_021844766.1 | XP_021700458.1 | 2 - 3 | 323 | 79 | ||
XM_021844767.1 | XP_021700459.1 | 2 - 3 | 209 | 79 |
