Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 14 - 15 | 4572 | 84 | |
XM_021844764.1 | XP_021700456.1 | 14 - 15 | 4684 | 84 | ||
XM_021844765.1 | XP_021700457.1 | 14 - 15 | 4528 | 84 | ||
XM_021844766.1 | XP_021700458.1 | 14 - 15 | 4629 | 84 | ||
XM_021844767.1 | XP_021700459.1 | 14 - 15 | 4515 | 84 | ||
XM_021844768.1 | XP_021700460.1 | 13 - 14 | 4400 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
