Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 12 - 13 | 3614 | 89 | |
XM_021844764.1 | XP_021700456.1 | 12 - 13 | 3726 | 89 | ||
XM_021844765.1 | XP_021700457.1 | 12 - 13 | 3570 | 89 | ||
XM_021844766.1 | XP_021700458.1 | 12 - 13 | 3671 | 89 | ||
XM_021844767.1 | XP_021700459.1 | 12 - 13 | 3557 | 89 | ||
XM_021844768.1 | XP_021700460.1 | 11 - 12 | 3442 | 89 |
