Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 11 - 12 | 3359 | 80 | |
XM_021844764.1 | XP_021700456.1 | 11 - 12 | 3471 | 80 | ||
XM_021844765.1 | XP_021700457.1 | 11 - 12 | 3315 | 80 | ||
XM_021844766.1 | XP_021700458.1 | 11 - 12 | 3416 | 80 | ||
XM_021844767.1 | XP_021700459.1 | 11 - 12 | 3302 | 80 | ||
XM_021844768.1 | XP_021700460.1 | 10 - 11 | 3187 | 80 |
