Assay Details
Gene Symbol: LOC5567934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843889.1 | XP_021699581.1 | 2 - 3 | 1344 | 74 | |
XM_021843898.1 | XP_021699590.1 | 2 - 3 | 1340 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
