Assay Details
Gene Symbol: LOC5576036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842628.1 | XP_021698320.1 | 7 - 8 | 2808 | 62 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5576036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842628.1 | XP_021698320.1 | 7 - 8 | 2808 | 62 |
Set Membership: |
![]() ![]() |