Assay Details
Gene Symbol: LOC5570582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849030.1 | XP_021704722.1 | 12 - 13 | 3140 | 61 | |
XM_021849031.1 | XP_021704723.1 | 12 - 13 | 2608 | 61 | ||
XM_021849032.1 | XP_021704724.1 | 11 - 12 | 3128 | 61 | ||
XM_021849033.1 | XP_021704725.1 | 11 - 12 | 3068 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
