Assay Details
Gene Symbol: LOC5570582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849030.1 | XP_021704722.1 | 14 - 15 | 3670 | 65 | |
XM_021849031.1 | XP_021704723.1 | 14 - 15 | 3138 | 65 | ||
XM_021849032.1 | XP_021704724.1 | 13 - 14 | 3658 | 65 | ||
XM_021849033.1 | XP_021704725.1 | 13 - 14 | 3598 | 65 |

More Information
Set Membership: |
![]() ![]() |
