Assay Details
Gene Symbol: LOC5570582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849030.1 | XP_021704722.1 | 1 - 2 | 864 | 81 | |
XM_021849032.1 | XP_021704724.1 | 1 - 2 | 864 | 81 | ||
XM_021849033.1 | XP_021704725.1 | 1 - 2 | 864 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849030.1, XM_021849032.1, XM_021849033.1 transcripts.
Set Membership: |
![]() ![]() |
