Assay Details
Gene Symbol: LOC5570582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849030.1 | XP_021704722.1 | 8 - 9 | 2700 | 66 | |
XM_021849031.1 | XP_021704723.1 | 8 - 9 | 2168 | 66 | ||
XM_021849032.1 | XP_021704724.1 | 8 - 9 | 2700 | 66 | ||
XM_021849033.1 | XP_021704725.1 | 7 - 8 | 2628 | 66 |

More Information
Set Membership: |
![]() ![]() |
