Assay Details
Gene Symbol: LOC5575163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846594.1 | XP_021702286.1 | 1 - 2 | 437 | 62 | |
XM_021846595.1 | XP_021702287.1 | 1 - 2 | 439 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
