Assay Details
Gene Symbol: LOC5578399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841942.1 | XP_021697634.1 | 1 - 2 | 1753 | 72 | |
XM_021841943.1 | XP_021697635.1 | 1 - 2 | 1753 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
