Assay Details
Gene Symbol: LOC5569921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840084.1 | XP_021695776.1 | 6 - 7 | 3664 | 87 | |
XM_021840090.1 | XP_021695782.1 | 6 - 7 | 3754 | 87 | ||
XM_021840093.1 | XP_021695785.1 | 6 - 7 | 3720 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
