Assay Details
Gene Symbol: LOC5569921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840084.1 | XP_021695776.1 | 5 - 6 | 3426 | 55 | |
XM_021840090.1 | XP_021695782.1 | 5 - 6 | 3516 | 55 | ||
XM_021840093.1 | XP_021695785.1 | 5 - 6 | 3482 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
