Assay Details
Gene Symbol: LOC5569921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840084.1 | XP_021695776.1 | 2 - 3 | 361 | 54 | |
XM_021840090.1 | XP_021695782.1 | 2 - 3 | 451 | 54 | ||
XM_021840093.1 | XP_021695785.1 | 2 - 3 | 417 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
