Assay Details
Gene Symbol: LOC110674772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839060.1 | XP_021694752.1 | 11 - 12 | 4176 | 66 | |
XM_021839061.1 | XP_021694753.1 | 10 - 11 | 4170 | 66 | ||
XM_021839062.1 | XP_021694754.1 | 10 - 11 | 3921 | 66 | ||
XM_021839063.1 | XP_021694755.1 | 11 - 12 | 3425 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
