Assay Details
Gene Symbol: LOC110674772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839060.1 | XP_021694752.1 | 6 - 7 | 2985 | 106 | |
XM_021839061.1 | XP_021694753.1 | 6 - 7 | 2985 | 106 | ||
XM_021839063.1 | XP_021694755.1 | 6 - 7 | 2234 | 106 |

More Information
Set Membership: |
![]() ![]() |
