Assay Details
Gene Symbol: LOC5570514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659152.2 | XP_001659202.1 | 1 - 2 | 158 | 134 | |
XM_021840732.1 | XP_021696424.1 | 1 - 2 | 159 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001659152.2, XM_021840732.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
