Assay Details
Gene Symbol: LOC5575216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850012.1 | XP_021705704.1 | 3 - 4 | 1460 | 100 | |
XM_021850013.1 | XP_021705705.1 | 3 - 4 | 1460 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021850012.1, XM_021850013.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
