Assay Details
Gene Symbol: LOC5575216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850012.1 | XP_021705704.1 | 5 - 6 | 2599 | 62 | |
XM_021850013.1 | XP_021705705.1 | 5 - 6 | 2599 | 62 | ||
GenBank mRNA | JN030894.1 | - | 2 - 3 | 658 | 62 | |
KC439534.1 | - | 2 - 3 | 1103 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
